Learn More
Thermo Scientific™ DyNAmo™ cDNA Synthesis Kit
Perform quantitative reverse transcription PCR (qRT-PCR) applications optimized for amplicons 100bp in length with Thermo Scientific™ DyNAmo™ cDNA Synthesis Kit.
Brand: Thermo Scientific™ F-470L
Code : A0
Additional Details : Weight : 1.71460kg
Description
Thermo Scientific™ DyNAmo™ cDNA Synthesis Kit is optimized for quantitative reverse transcription PCR (qRT-PCR) applications, where amplicons are typically 100 bp in length.
The DyNAmo cDNA Synthesis Kit includes all necessary reagents for cDNA synthesis from various RNA sources. Total RNA, messenger RNA, viral RNA or in vitro transcribed RNA can all be used as a template for reverse transcription.- Specifically optimized for quantitative RT-PCR
- Broad dynamic range: up to 1μg of RNA
- Reliable results with different RNA sources: mRNA, total RNA, viral RNA and in vitro transcribed RNA
- Flexible: applicable with all priming options for cDNA synthesis
- Easy to use: short protocol, 2x mix
Includes:
M-MuLV RNase H+ reverse transcriptase (includes RNase inhibitor), 2x RT buffer (includes dNTP mix and 10 mM MgCl2), Random hexamers (300 ng/μL), Oligo(dT)15 primer (100 ng/μL).
Recommended for:
cDNA synthesis for qRT-PCR
Specifications
M-MuLV RNase H+ reverse transcriptase (includes RNase inhibitor), 2x RT buffer (includes dNTP mix and 10 mM MgCl2), Random hexamers (300 ng/µL), Oligo(dT)15 primer (100 ng/µL). Store at -20°C. | |
cDNA synthesis for qRT-PCR | |
20 reactions | |
cDNA Synthesis |