Learn More
DyNAmo ColorFlash DyNamo ColorFlash Probe qPCR Kit
Offers equal performance to the DyNAmo Flash Probe qPCR Kit. In addition, it incorporates an innovative multicolor system that ensures correct pipetting.
152.00 € - 2360.00 €
Specifications
Detection Method | Primer-probe |
---|---|
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Tbr DNA polymerase |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10401358
|
Thermo Scientific™
F456L |
500 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10738908
|
Thermo Scientific™
F456S |
100 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10164039
|
Thermo Scientific™
F456XL |
2500 Reactions |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
DyNAmo ColorFlash Probe qPCR Kit offers equal performance to the DyNAmo Flash Probe qPCR Kit. In addition, it incorporates an innovative multicolor system that ensures correct pipetting. The 2X master mix contains a blue dye, and a separate sample buffer contains a yellow dye. The qPCR reaction mix containing both components is green. Using this patent-pending multicolor system, pipetting of both the master mix and the sample can be easily monitored. This significantly decreases the risk for pipetting errors during reaction set up, especially when using white reaction vessels. The dyes do not affect the specificity or sensitivity of qPCR assays.
- Minimized risk of pipetting errors during reaction set up
- Especially beneficial when using white reaction vessels
- Extremely fast protocols due to combined annealing and extension step of only 15 sec.
- Specific and sensitive detection of a wide range of template concentrations
- dUTP included in the 2X master mix allows the use of UNG for prevention of carry-over contamination
Specifications
Primer-probe | |
High | |
Tbr DNA polymerase | |
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Stratagene Mx4000, StepOne™ System, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, BioRad MyiQ, BioRad iQ5, BioRad iCycler iQ, Cepheid SmartCycler, Stratagene Mx3005P | |
DyNAmo Flash | |
Gene Expression | |
DNA, dsDNA |
Tube | |
qPCR | |
Fast | |
DyNAmo ColorFlash Probe qPCR Kit | |
Real Time PCR SYBR Master Mix | |
2X (DyNAmo ColorFlash qPCR Mix), 50X (ROX Dye), 40X (Sample Buffer Solution with Yellow Dye) |
For Research Use Only. Not for use in diagnostic procedures.