Learn More
DyNAmo ColorFlash SYBR™ DyNAmo ColorFlash SYBR Green qPCR Kit
Offers equal performance to the DyNAmo Flash SYBR Green qPCR Kit. In addition, it incorporates an innovative multi-color system that ensures correct pipetting.
Brand: DyNAmo ColorFlash SYBR™ F416S
Description
The 2X master mix contains a blue dye, and a separate sample buffer contains a yellow dye. The qPCR reaction mix containing both components is green. Using this patent-pending multicolor system, pipetting of both the master mix and the sample can be easily monitored. This significantly decreases the risk for pipetting errors during reaction setup, especially when using white reaction vessels. The dyes do not affect the specificity or sensitivity of qPCR assays.
- Colored reagents minimize the risk of pipetting errors during reaction set up, especially beneficial when using white reaction vessels
- Extremely fast protocols due to combined annealing and extension step of only 15 sec.
- Specific and sensitive detection of a wide range of template concentrations
- dUTP included in the 2X master mix allows the use of UNG for prevention of carry-over contamination
Specifications
Contains: • 1 x 1 mL DyNAmo ColorFlash SYBR Green qPCR Mix (2X) • 1 x 50 μL 50X ROX dye, 25μM • 1 x 200 μL 40x sample buffer solution with yellow dye Store at -20°C |
|
Tube | |
qPCR | |
Fast | |
DyNAmo ColorFlash SYBR Green qPCR Kit | |
Real Time PCR SYBR Master Mix | |
Gene Expression | |
100 reactions | |
1 x 1 mL |
SYBR | |
High | |
Tbr DNA polymerase | |
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT Fast System, 7900HT System, BioRad CFX96™, QuantStudio™, ViiA™ 7 System, Bio-Rad CFX384 Touch™, Roche LightCycler 480, 5700 Sequence Detection System, StepOnePlus™ System, Eppendorf MasterCycler RealPlex, Corbett RotorGene, StepOne™ System, Stratagene Mx4000, Stratagene Mx3005P, Stratagene Mx3000P, MJ Opticon, MJ Chromo4, Cepheid SmartCycler | |
DyNAmo Flash | |
100 Reactions of 20 μL, 40 Reactions of 50 μL | |
2X (DyNAmo ColorFlash SYBR Green qPCR), 50X (ROX Dye), 40X (Sample Buffer Solution with Yellow Dye) | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.