missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin (CDH1) / CD324 (Intercellular Junction Marker) Mouse, Clone: rCDH1/1525, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | E-Cadherin (CDH1) / CD324 (Intercellular Junction Marker) |
---|---|
Clone | rCDH1/1525 |
Concentration | 200μg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15961328
|
EnquireBio
999-MSM7-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951328
|
EnquireBio
999-MSM7-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
E-Cadherin (CDH1) / CD324 (Intercellular Junction Marker) | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human E-Cadherin protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
120-80kDa (Mature); 135kDa (Precursor) |
rCDH1/1525 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
999 | |
IgG1 κ | |
Protein A/G purified from Bioreactor concentrate. | |
Cell Surface | |
MCF-7, LS174T, Raji, HT29, SK-BR3 cells. Prostate or Colon carcinomas. |