missing translation for 'onlineSavingsMsg'
Learn More
Learn More
E-Cadherin (CDH1) / CD324 (Intercellular Junction Marker) Mouse, Clone: rCDH1/1525, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 999-MSM7-P1
Additional Details : Weight : 0.01000kg
Specifications
E-Cadherin (CDH1) / CD324 (Intercellular Junction Marker) | |
Cell Surface | |
rCDH1/1525 | |
Unconjugated | |
Mouse | |
120-80kDa (Mature); 135kDa (Precursor) | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
999 | |
IgG1 κ |
Immunohistochemistry, Western Blot | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant full-length human E-Cadherin protein | |
MCF-7, LS174T, Raji, HT29, SK-BR3 cells. Prostate or Colon carcinomas. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |