Learn More
IBA Lifesciences pASG-IBA E. coli Expression Vector with No Secretion
Regulate the expression of proteins with a vector that carries the inducible tetracycline promoter/operator. E. coli Expression Vector with No Secretion allows for the expression of proteins in E. coli.
220.00 € - 286.00 €
Specifications
Inducing Agent Inducible with anhydrotetracycline
Marker Gene Ampicillin
Promoter Tet
Product Type Vector
Content And Storage Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage
View More Specs
Products 22
Product Code Brand Vector Price Quantity & Availability  
Product Code Brand Vector Price Quantity & Availability  
15603758
View Documents Promotion Details
IBA Lifesciences
5-4164-001
pASG-IBA164
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15613758
View Documents Promotion Details
IBA Lifesciences
5-4167-001
pASG-IBA167
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15623748
View Documents Promotion Details
IBA Lifesciences
5-4103-001
pASG-IBA103
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15623758
View Documents Promotion Details
IBA Lifesciences
5-4168-001
pASG-IBA168
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15633728
View Documents Promotion Details
IBA Lifesciences
5-4000-001
pASG-IBAwt1
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15633738
View Documents Promotion Details
IBA Lifesciences
5-4043-001
pASG-IBA43
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15643748
View Documents Promotion Details
IBA Lifesciences
5-4105-001
pASG-IBA105
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15653738
View Documents Promotion Details
IBA Lifesciences
5-4045-001
pASG-IBA45
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15653748
View Documents Promotion Details
IBA Lifesciences
5-4123-001
pASG-IBA123
286.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15663728
View Documents Promotion Details
IBA Lifesciences
5-4003-001
pASG-IBA3
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15663738
View Documents Promotion Details
IBA Lifesciences
5-4062-001
pASG-IBA62
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15663748
View Documents Promotion Details
IBA Lifesciences
5-4143-001
pASG-IBA143
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15683728
View Documents Promotion Details
IBA Lifesciences
5-4005-001
pASG-IBA5
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15683738
View Documents Promotion Details
IBA Lifesciences
5-4064-001
pASG-IBA64
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15683748
View Documents Promotion Details
IBA Lifesciences
5-4145-001
pASG-IBA145
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15693728
View Documents Promotion Details
IBA Lifesciences
5-4023-001
pASG-IBA23
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15693748
View Documents Promotion Details
IBA Lifesciences
5-4162-001
pASG-IBA162
266.00 €
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
15623738
View Documents Promotion Details
IBA Lifesciences
5-4035-001
pASG-IBA35 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15613738
View Documents Promotion Details
IBA Lifesciences
5-4033-001
pASG-IBA33 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15673738
View Documents Promotion Details
IBA Lifesciences
5-4063-001
pASG-IBA63 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15693738
View Documents Promotion Details
IBA Lifesciences
5-4065-001
pASG-IBA65 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15603738
View Documents Promotion Details
IBA Lifesciences
5-4025-001
pASG-IBA25 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Regulate the expression of proteins with a vector that carries the inducible tetracycline promoter/operator. IBA Lifesciences GmbH E. coli Expression Vector with No Secretion allows for the expression of proteins in E.coli.
Specifications
Specifications
Inducible with anhydrotetracycline
Tet
Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage
Ampicillin
Vector
5 μg