missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Eco AluRack for Upright Freezer, for Microtiterplates, 65 mm Shelf Height
Eco AluRack for upright freezers is designed to hold microtiterplates.
71.55 € - 148.00 €
Specifications
Material | Aluminum |
---|---|
For Use With (Equipment) | Upright Freezers |
Height (Metric) | 65 mm |
Product Code | Brand | Holds | Array | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Array | Price | Quantity & Availability | ||||
12418220
|
Tenak™
TE15430 |
Microtiterplates (8) | 2 x 4 |
71.55 €
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
12428220
|
Tenak™
TE15432 |
Microtiterplates (12) | 3 x 4 |
83.50 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
12438220
|
Tenak™
TE15434 |
Microtiterplates (16) | 4 x 4 |
95.40 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
12438240
|
Tenak™
TE15500 |
Microtiterplates (10) | 2 x 5 |
81.20 €
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
12448240
|
Tenak™
TE15502 |
Microtiterplates (15) | 3 x 5 |
93.15 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
12448260
|
Tenak™
TE15550 |
Microtiterplates (12) | 2 x 6 |
90.85 €
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
12458240
|
Tenak™
TE15504 |
Microtiterplates (20) | 4 x 5 |
108.00 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
12458260
|
Tenak™
TE15552 |
Microtiterplates (18) | 3 x 6 |
106.00 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
12458280
|
Tenak™
TE15600 |
Microtiterplates (14) | 2 x 7 |
99.95 €
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
12468240
|
Tenak™
TE15506 |
Microtiterplates (25) | 5 x 5 |
120.00 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
12468260
|
Tenak™
TE15554 |
Microtiterplates (24) | 4 x 6 |
120.00 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
12468280
|
Tenak™
TE15602 |
Microtiterplates (21) | 3 x 7 |
115.00 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
12478240
|
Tenak™
TE15508 |
Microtiterplates (30) | 6 x 5 |
134.00 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
12478260
|
Tenak™
TE15556 |
Microtiterplates (30) | 5 x 6 |
134.00 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
12478280
|
Tenak™
TE15604 |
Microtiterplates (28) | 4 x 7 |
129.00 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
12488260
|
Tenak™
TE15558 |
Microtiterplates (36) | 6 x 6 |
148.00 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
12488280
|
Tenak™
TE15606 |
Microtiterplates (35) | 5 x 7 |
144.00 €
Each |
Estimated Shipment: 23-09-2024
Log in to see stock. |
||||
Description
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
Aluminum | |
Upright Freezers | |
65 mm |