missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Elastase Inhibitor II
A potent irreversible inhibitor of human neutrophil elastase
POA
Specifications
Description | White |
---|---|
Content And Storage | -30°C to -10°C |
For Use With (Application) | A potent irreversible inhibitor of human neutrophil elastase |
Molecular Formula | C20H31ClN4O7 |
Formula Weight | 474.90g/mol |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
White | |
A potent irreversible inhibitor of human neutrophil elastase | |
474.90g/mol | |
Moisture Sensitive | |
Signal Transduction Reagent |
-30°C to -10°C | |
C20H31ClN4O7 | |
HNE Inhibitor; MeOSuc-AAPA-CMK | |
Soluble in methanol or DMSO | |
Solid |