Learn More
SCAT Connectors for HPLC
Safe connection of tubes of different sizes: directly and without technical aids. For safe collection of liquid HPLC waste, and many other applications in the laboratory!
Brand: SCAT 107808
Code : NEW
Additional Details : Weight : 0.10000kg
Description
SCAT™ connectors are available in the following versions:
- Conical connectors
- Y-connectors
- Tube connectors
- Capillary connectors
Specifications
Tube Connector | |
Polypropylene | |
HPLC/UHPLC Systems |
6.4 to 8 mm | |
Tubing |