Learn More
Veolia Water Systems Elgas™ Cartridge
Deionization cartridge compatible with systems that remove ionic contaminants to achieve specified water quality.
Brand: Veolia Water Systems CODILC0141
Code : 04
Additional Details : Weight : 2.18700kg
Description
- Removes ionic contaminants to achieve specified water quality
- Compatible with MEDICA-D 7/15, MEDICA-R 7/15, MEDICA-S 7/15,
- PURELAB Option-R 7/15, PURELAB Option-S 7/15
- Maximum operating temperature 95 degrees F