missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Amicon™ Ultra-2.0 Centrifugal Filter Units
Reverse spin capability provides consistent recoveries
242.00 € - 267.00 €
Specifications
Material (Membrane) | Regenerated Cellulose |
---|---|
Filtration Area | 1 cm2 |
For Use With (Application) | Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms; Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC: Desalting and buffer exchange |
Includes | Two microcentrifuge tubes |
Max. Force | 14,000xG for concentration spin; 1000xG for reverse spin |
Includes: Two microcentrifuge tubes
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Price | Quantity & Availability | |||||
11592134
|
Merck
UFC201024 |
10 kDa |
245.00 €
Pack of 24 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||||
11512144
|
Merck
UFC203024 |
30 kDa |
267.00 €
Pack of 24 |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
11582134
|
Merck
UFC200324 |
3 kDa |
242.00 €
Pack of 24 |
In Stock
Log in to see stock. |
|||||
13105096
|
Merck
UFC210024 |
100 kDa |
267.00 €
Pack of 24 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
Description
- Concentrate 2mL down to 15 – 70μL
- True engineered dead stop provides consistent recovery without spinning to dryness
- Reverse spin aids in sample recovery, especially with small, dilute samples
- Vertical membrane design aligns with the filtrate for less clogging and faster filtering
- Ultracel low-binding membrane combines with the innovative vertical housing to provide fast sample processing without sacrificing recovery
- Fast processing, with 10- to 30-minute spin times
Applications:
- Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
- Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC
- Desalting and buffer exchange
Specifications
Regenerated Cellulose | |
Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms; Purification of macromolecular components found in tissue culture extracts or cell lysates, removal of primer, linkers, or macromolecular labels from a reaction mix, and protein removal prior to HPLC: Desalting and buffer exchange | |
14,000xG for concentration spin; 1000xG for reverse spin | |
2.0 mL | |
4.99 cm |
1 cm2 | |
Two microcentrifuge tubes | |
24/Pk. | |
0.196 in. |