missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Centriprep™ Centrifugal Filter Units
For processing high-solute samples
358.00 € - 1635.00 €
Specifications
Disposable | Single-use |
---|---|
Material (Membrane) | Ultracel-YM Regenerated Cellulose |
Sterility | Non-sterile |
Filtration Area | 2.84 cm2 |
Housing Material | Regenerated Cellulose (RC) |
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | MWCO | Quantity | Price | Quantity & Availability | ||||
10733121
|
Merck
4304 |
10 kDa | 24Pack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10278490
|
Merck
4303 |
3 kDa | 96Pack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10626551
|
Merck
4311 |
50 kDa | 96Pack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10509412
|
Merck
4307 |
30 kDa | 96Pack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
For processing high-solute samples
Centriprep™ centrifugal filters are disposable ultrafiltration devices used for purifying, concentrating, and desalting biological samples in the 2–15 mL volume range. These complete, ready-to-use ultrafiltration devices are designed for operation in most centrifuges that can accommodate 50 mL centrifuge tubes. They are easy to use and offer a high flow rate. Centriprep™ devices consist of a sample container with a twist-lock cap, a filtrate collector containing a low-adsorptive, Ultracel™ regenerated cellulose membrane and an air-seal cap for sample isolation.Features & Benefits:
Applications:Concentration and Purification of Particle-laden Solutions, Solutions with High Solute Concentration, Separation of Low MW Solutes from Fermentation Broths, Cell Culture Media and Lysates
Specifications
Single-use | |
Non-sterile | |
Regenerated Cellulose (RC) | |
12 to 15 mL |
Ultracel-YM Regenerated Cellulose | |
2.84 cm2 | |
Centrifugal Filters | |
135 mm |