Learn More
Merck Durapore™ Membrane Filters, 0.22 μm
Recommended when maximum recovery is important. Durapore™ PVDF Membranes: 0.22μ Pore Size provide high flow rates and throughput, low extractables, broad chemical compatibility and low protein binding. Available in diameters ranging from 13mm to 142mm.
189.00 € - 1190.00 €
Specifications
Autoclavable | Not Autoclavable |
---|---|
Color | White |
Material | PVDF |
Pore Size | 0.22 μm |
Sterility | Non-sterile |
Product Code | Brand | Diameter (Metric) Filter | Bubble Point | Filter Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Filter | Bubble Point | Filter Type | Price | Quantity & Availability | |||
10752901
|
Merck
GVWP04700 |
47 mm | 50 psi | Screen Filter |
487.00 €
Pack of 100 |
In Stock
Log in to see stock. |
|||
10742901
|
Merck
GVWP02500 |
25 mm | 50 psi | Screen Filter |
231.00 €
Pack of 100 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||
10074500
|
Merck
GVWP01300 |
13 mm | 50 psi | Screen Filter |
189.00 €
Pack of 100 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||
10732731
|
Merck
GVWP14250 |
142 mm | 50 psi | Screen Filter |
750.00 €
Pack of 50 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||
10146710
|
Merck
GVWP09050 |
90 mm | 50 psi | Screen Filter |
687.00 €
Pack of 50 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||
10656741
|
Merck
GVHP04700 |
47 mm | ≥1.24 bar, air with Methanol | Screen Filter |
459.00 €
Pack of 100 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||
10646741
|
Merck
GVHP02500 |
25 mm | ≥1.24 bar, air with Methanol | Screen Filter |
321.00 €
Pack of 100 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||
10646551
|
Merck
GVHP01300 |
13 mm | ≥1.24 bar, air with Methanol | Screen Filter |
259.00 €
Pack of 100 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||
10696161
|
Merck
GVHP09050 |
90 mm | ≥1.24 bar, air with Methanol | Screen Filter |
610.00 €
Pack of 50 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||
10742151
|
Merck
GVHP14250 |
142 mm | ≥1.24 bar, air with Methanol | Screen Filter |
945.00 €
Pack of 50 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||
15869542
|
Merck
GVWP2932A |
293 mm | 53≤x ≤60 psi | Screen Filter |
890.00 €
Pack of 25 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||
10479830
|
Merck
GVWP10050 |
100 mm | ≥3.45 bar, air with water | Screen Filter |
631.00 €
Pack of 50 |
Estimated Shipment: 08-07-2024
Log in to see stock. |
|||
15858077
|
Merck
GVWP07625 |
76 mm | ≥50 psi, air with water | Screen Filter |
191.00 €
Pack of 25 |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||
10606171
|
Merck
GVWP00010 |
304 mm | ≥3.45 bar, air with water | Screen filter, 1 roll x 10 ft. |
1190.00 €
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||
10194073
|
Merck
GVHP00010 |
304 mm | ≥1.24 bar, air with Methanol | Screen filter, 1 roll x 10 ft. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Recommended when maximum recovery is important. Durapore™ PVDF Membranes: 0.22μ Pore Size provide high flow rates and throughput, low extractables, broad chemical compatibility and low protein binding. Available in diameters ranging from 13mm to 142mm.
Durapore™ membranes provide high flow rates and throughput, low extractables and broad chemical compatibility. Hydrophlic Durapore™ membrane binds far less protein than nylon, nitrocellulose, or PTFE membranes.- Features & Benefits
Specifications
Not Autoclavable | |
PVDF | |
Non-sterile |
White | |
0.22 μm | |
Filter Membranes |