missing translation for 'onlineSavingsMsg'
Learn More

Merck Express™ PLUS Membrane Filters

Plain white polyethersulfone Express™ PLUS Membrane Filters provide ultrafast filtration for a variety of applications.

Brand:  Merck HPWP01300

 View more versions of this product

Product Code. 10022433

  • 210.00 € / Pack of 100
Only null left
Estimated Shipment: 01-09-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications

The Millipore Express™ PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.

Features and Benefit:
  • Ultra fast filtration and high throughput shorten process time
  • Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications
  • Applications:Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
Reusable
Disc
0.45 μm
23 (at 690 Mbar (10 psi))
Hydrophilic
Hydrophilic
13 mm
White
Screen Filter
PES
Non-sterile
Provide ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This membrane is also used in many ready-to-use sterile filtration devices.
Filter Membranes
0.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.