missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Chemicals EZ-Fit™ Filtration Unit
For bioburden testing of liquid samples like water, process samples, or final products
POA
Specifications
Material (Membrane) | MCE |
---|---|
Pore Size | 0.45 μm |
Sterility | Sterile |
No. per Pack | 48 |
Quantity | 48/Pk. |
Description
- Designed for optimizing and securing laboratory workflow to provide time-saving and reliable microbiological results
- Comprise of 100mL or 250mL funnels
- Black or white gridded membranes
- After filtration of sample, membrane can be transferred and cultured on agar plate
- Sterilized and disposable
- Units are either individually bagged or packaged in bulk
Conform to international standards (EP/USP) and comply with water testing regulations.
Specifications
MCE | |
Sterile | |
48/Pk. |
0.45 μm | |
48 |