missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories for XX6702500 FilterJet Solvent Dispenser

Accessories for filter holders for sample preparation.

Brand:  Merck XX6702502

Code : 72

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10682822

  • 152.00 € / Pack of 4
Estimated Shipment: 10-05-2024
to see stock.
Add to basket

Description

Description

Accessories for filter holders for sample preparation.

  • Made of PTFE
  • Connectors: 1/4 in. NPTF elbow fittings for inlet and outlet (labeled) and 1/4 in. NPTF for pressure gauge and vent/relief valve attachment

Specifications

Specifications

Not Autoclavable
Valve Seals
PTFE
7 bar (100 psi)
1/4 in. NPTF elbow fittings for inlet and outlet (labeled); 1/4 in. NPTF for pressure gauge and vent/relief valve attachment
Filterjet Solvent Dispenser
Non-sterile
Valve Seals
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers