Learn More
Merck Millipore Hydrophobic 6mm Edge Membranes
For microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations
Brand: Merck Millipore GSEP0470W
Code : 71
Additional Details : Weight : 0.02000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
- Nonwetting edge prevents solution intrusion under holder sealing rings
- Hydrophobic edge membranes are standard MF-Type (mixed cellulose ester) filters
- 47mm diameter
Specifications
White | |
4.7 cm | |
Plain | |
Mixed Cellulose Ester | |
Non-sterile |
1.85 in. | |
Microbiological and sterility testing of antibiotics and bacteriostatic drugs, as well as for filling machine operations, filling machine operations, sterility testing | |
Sterile pack: 10 resealable envelopes containing 10 filters and 10 pads | |
0.22 μm |