missing translation for 'onlineSavingsMsg'
Learn More

Merck MF-Millipore™ Membrane Filter, 0.45 μm, Gridded

Mixed cellulose esters (MCE) membrane, hydrophilic

Brand:  Merck HABG04700

Code : 71

Additional Details : CAS Number : 9004-70-0 Weight : 0.14664kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 10054650

  • 282.61 € / Pack of 100

Estimated Shipment: 16-12-2021
Add to basket

Description

Description

Mixed cellulose esters (MCE) membrane, hydrophilic

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.

Features & Benefits:

  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications

Specifications

0.45 μm
Non-sterile
Gridded
47 mm
150 µm
Not Autoclavable
60 mL/min x cm2
100Pack
Hydrophilic
Mixed Cellulose Esters (MCE) (Nitrate and Acetate)
Filter Membranes
Black
47 mm
Hydrophilic
≥2.2 bar, air with water
4 L/min/ cm2
100/Pk.
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MCE .45UM BK GD 47MM 100/PK

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use to extinguish.

SDS
Documents
Certificates
Special Offers

Special Offers