Learn More
Merck Millex™ Nonsterile Syringe Filters with PE Housing - Mitex
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Brand: Merck SLLS025NS
313.67 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Code : 71
Additional Details : Weight : 0.01000kg
Description
- Hydrophobic PTFE Membrane
- Compatible with more aggressive organic solutions and high temperature conditions
†With tube outlet, for filtration directly into small receiving vessels.
Specifications
Mitex | |
Polyethylene | |
25 mm |
5 μm | |
Syringe Filter |