missing translation for 'onlineSavingsMsg'
Learn More

Merck Millex™-GP Syringe Filter, PES, 0.22 μm

Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses

Brand:  Merck SLGP033NS

Code : 71

Additional Details : Weight : 0.21620kg

Product Code. 10747984

  • 249.00 € / Pack of 50
Estimated Shipment: 25-06-2024
to see stock.
Add to basket

Description

Description

Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses

Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:
  • Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
  • Available in 0.22 μm and 0.45 μm pore sizes and two diameters to suit your application needs
  • Low protein binding to minimize interaction with your sample and maximize recovery
  • Polypropylene housing, offering a low level of extractables and broad chemical compatibility

Applications: Buffer Filtration, Clarification of Aqueous Solutions

Specifications

Specifications

Single-use
PES
Non-sterile
Analytical sample prep
Male Luer slip
50Pack
<200 mL
33 mm
Millipore Express PLUS (PES)
0.22 μm
4.5 cm2
PP
Millex™
Syringe and Syringeless Filter
33 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers