Learn More
Merck Medical Millex™-GS Syringe Filter Unit, 0.22 μm
Ready-to-use, syringe-driven units designed for sterilizing and clarifying sterile solutions. Sterilized by EtO.
Brand: Merck SLGSM33SS
Code : NEW
Additional Details : Weight : 0.38700kg
Description
Ready-to-use, syringe-driven units designed for sterilizing and clarifying sterile solutions.
Available in 4, 13, 25, 33, and 50 mm diameters with a variety of membranes, Millex™ sterile syringe filters are ideal for sterilizing organic solvents, aqueous solutions or air/gas. Many sample preparation methods specify Millex™ filters because of their unsurpassed quality and consistency. Millex™ filters not only feature minimal hold-up volume and faster flow rates, but they also withstand higher operating pressures and are chemically compatible with a range of samples and solvents to minimize leaching of extractable impurities. Millex™ filters are produced in a controlled, automated environment with the highest quality standards. Sterile devices come with a certificate of quality, with select devices marked for medical applications.Features and Benefits:
- Tailored to a variety of applications based on membrane type, housing and validation rating
- Medical Device-rated devices in some countries including vented devices for direct patient care
- Specialized filter units to protect hemodialysis transducers from blood and moisture and for vacuum line protection
- Low extractables and low absorption membranes for minimum sample losses
- Faster flow rates and higher operating pressures for high performance needs
Applications: Sterilization of Aqueous Solutions, Organic Solvents and Gases, Filtration of High Value Protein or Small Molecule Components, Select Medical Device Applications (Some Including Direct Patient Care Depending on Country/Region)
Specifications
Not Autoclavable | |
Blue | |
Mixed Cellulose Esters (MCE) | |
Sterile | |
<100 μL | |
Female Luer Lok | |
145 psi | |
113°F | |
113°F | |
Modified Acrylic | |
200 mL |
Meets USP Class VI Biological tests for plastics. All 33 mm filters are CE-marked and suitable for both medical and laboratory applications outside the U.S. | |
Single-use | |
0.22 μm | |
Clarification of water, buffers or aqueous solutions | |
Mixed Cellulose Esters (MCE) | |
Male Luer Slip | |
50Pack | |
45°C | |
45°C | |
Syringe Filter (Medical) | |
33 mm |