missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millipore Millicup™ Self-Contained, Disposable, Vacuum Bottle-Top Filter for HPLC
Non sterile Universal bottletop vacuum filter
POA
Specifications
Type | Bottle Top Filter |
---|---|
For Use With (Application) | HPLC |
Height (English) | 4.7 in. |
Height (Metric) | 12 cm |
Pore Size | 0.45 μm |
Description
The Millicup vacuum-driven bottletop filter unit is a disposable device that provides a fast, effective, and safe means to filter mobile phases and running buffers used in instrumentation analyses (for example, HPLC). The Millicup vacuum-driven bottletop filter unit eliminates the risk of contaminant carry-over from solvent to solvent. Unlike reusable filter holders that must be washed and assembled before each use, Millicup units require no assembly or clean-up. If accidentally dropped, the Millicup vacuum-driven bottletop filter unit will not break, ensuring a safe environment in which to filter solvents and other hazardous solutions.Specifications
Bottle Top Filter | |
4.7 in. | |
0.45 μm |
HPLC | |
12 cm |