Learn More
Merck MultiScreenHTS 96-Well Filter Plates
For filtration-based solubility determination. MultiScreenHTS 96-Well Filter Plates are specifically developed for use with automation.
Brand: Merck MSIPS4510
Code : 73
Additional Details : Weight : 0.84600kg
Description
For filtration-based solubility determination. MultiScreenHTS 96-Well Filter Plates are specifically developed for use with automation.
- High Throughput
- Meet ANSI/SBS 2004 plate specifications
- Protects underdrain, eliminates surface contact with individual well ‘drip directors’
- Plastic skirt enables stacking directly with collection plates
Applications
- Receptor-ligand binding; general sample preparation; Elispot; enzyme activity; bead-based assays; whole organism visualisation
- Radioisotope detection
Specifications
ANSI, SBS | |
Hydrophobic PVDF | |
0.45 μm | |
ABS Plastic | |
Sterile | |
ELISPOT assays |
Standard | |
8 x 12 | |
Clear | |
96 | |
0.28cm2 | |
10/Pk. |