missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreenHTS 96-Well Filter Plates
Non-sterile, clear 96-well filter plate with 0.45μm pore size Hydrophobic PVDF membrane for protein interaction assays, Avidin-biotin Linkages, DNA-binding proteins.
Brand: Merck MSIPN4510
Code : 73
Additional Details : Weight : 0.10000kg
Specifications
ANSI, SBS | |
Hydrophobic PVDF | |
0.45 μm | |
Clear | |
96 | |
0.28 cm2 | |
10/Pk. |
Standard | |
8 x 12 | |
Low Binding | |
Acrylic | |
Non-sterile | |
Protein interaction assays, avidin biotin linkages, DNA binding proteins |