missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™HTS 96-Well Filter Plates with Negatively Charged Membrane

Binds positive charge—ideal for kinase assays with peptide substrates and enzyme assays

Brand:  Merck MSPHN6B10

Code : 73

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10245703

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Two-piece plate design eliminates surface contact with individual well “drip directors,” prevents crosstalk between wells, allows membrane access, and provides easy, contamination-free stacking and improved seal with vacuum manifolds
  • Extended skirt enables stacking directly with collection plates, protects recessed underdrain, provides ample surface for barcode labels, and improves vacuum seal
  • Isolated well design and individually sealed filter discs eliminate potential for reagent flow from well to well
  • Meet ANSI™/SBS 2004 plate specifications
  • Standardized dimensions allow full compatibility with automated gripper arms, stackers, barcode labels, plate washers, and plate readers; aligned, rigid sidewalls permit easy handling by robotic gripper arms
  • Opaque plastic to prevents light transfer between wells during direct scintillation counting

Specifications

Specifications

ANSI, SBS
Negatively Charged Phosphocellulose
0.65 μm
MultiScreenHTS-PH Plate
0.05 to 0.25 mL
Non-sterile
Protein kinase assays with peptide substrates, enzyme assays, binds positive charge
10/Pk.
1.28 cm
1.44 cm
Standard
8 x 12
Opaque
Barex
96
0.26cm2
10
0.5 x 3.36 x 0.56 in. (1.28 x 8.55 x 1.44 cm)
8.55 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers