missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Pellicon™ XL Devices
Small-volume tangential flow filter device
935.00 € - 975.00 €
Specifications
Format | Cassette |
---|---|
Material | Polypropylene |
Pore Size | 100 μm |
Filtration Area | 50 cm2 |
For Use With (Application) | Protein purification, analytical sample preparation |
Includes: Operating instructions and Certificate of Quality
Product Code | Brand | Filter Type | pH Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Type | pH Range | Price | Quantity & Availability | ||||
11943926
|
Merck
PXB005A50 |
Ultrafiltration Biomax-5 | 1 to 14 |
975.00 €
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
10184739
|
Merck
PXB010A50 |
Ultrafiltration Biomax-10 | 1 to 14 |
935.00 €
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
12170212
|
Merck
PXB030A50 |
Ultrafiltration Biomax-30 | 1 to 14 |
935.00 €
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
11913916
|
Merck
PXB050A50 |
Ultrafiltration Biomax-50 | 1 to 14 |
935.00 €
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
13197494
|
Merck
PXC030C50 |
Ultrafiltration Ultracel-30 PLCTK | 2 to 21 |
935.00 €
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
12801666
|
Merck
PXC010C50 |
Ultrafiltration Ultracel-10 PLCGC | 2 to 21 |
935.00 €
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
Description
Designed for process development and sample preparation, with an optimal processing volume range of 50 to 1000mL.
- True linear scalability with same flow path, channel length, and channel height as other established Pellicon products
- Can operate with peristaltic pump or Labscale™ TFF system Robust thermoplastic (polypropylene) construction
- Controlled channel height
- Choice of Biomax™ PES (polyethersulfone) or Ultracel™ PLC (composite regenerated cellulose) membranes
Concentration and desalting of proteins, buffer exchange, depyrogenation, clarification, and preparation of material for clinical trials.
Order Info
Packaged one per box.
Specifications
Cassette | |
100 μm | |
Protein purification, analytical sample preparation | |
80psi at 50°C | |
0.03 to 0.05L/min. | |
0.5 oz. | |
15 mL | |
7.4 in. | |
1.2 in. |
Polypropylene | |
50 cm2 | |
Operating instructions and certificate of quality | |
1 Pack | |
Pellicon XL Ultrafiltration Device | |
3.2 mL | |
7.4 x 1.2 in. (18.8 x 3cm) | |
18.8 cm | |
3 cm |