Learn More
Merck Millipore Petri Dishes and Petri-Pad™ Dishes
Available with or without absorbent pads
534.00 €
Requires: When using with liquid medium, add sterile absorbent pads (order Cat. No. AP10 045 S0, 47mm Sterile Pads; or AP10 045 S1, Millipore Pad Dispenser with Sterile Pads)
| Product Code | Brand | dimType | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | dimType | Quantity | Price | Quantity & Availability | ||||
|
11993856
|
Merck Millipore
PD20047S5 |
534.00 €
Pack of 600 |
Estimated Shipment: 07-08-2026
Log in to see stock. |
||||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
Available with or without absorbent pads
Petri Dishes
- 47mm sterilized Petri dishes
- Use with solid agar medium
- Dishes are made of molded polystyrene
- Presterilized disposable 47mm dishes preloaded with sterilized absorbent pads
- Eliminate labor and cost of hand-loading pads and minimize contamination
- Pads are pure cellulose