Learn More
Merck Replacement Parts for Stainless-Steel 13 and 25mm Filter Holders
Use to ultraclean or sterilize small volumes of liquids dispensed by syringe
66.60 € - 294.00 €
Product Code | Brand | Material | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Includes | Price | Quantity & Availability | ||||
10652632
|
Merck
XX3001202 |
PTFE | Flat gasket |
127.00 €
Pack of 10 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
10273871
|
Merck
XX3001210 |
Stainless Steel | Support screen |
66.60 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
10492511
|
Merck
XX30025RK |
Stainless Steel | Two PTFE gaskets, two O-rings, and two support screens |
294.00 €
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
Description
Swinnex™ parts kit contains two PTFE gaskets, two O-rings, and two support screens.
Specifications
Stainless-Steel Filter Holder Replacement Parts |