Learn More
Merck Accessories and Replacement Parts In-Line Stainless Steel Filter Holders 47 mm
Accessories for filter holders for sample preparation.
Brand:  Merck XX4304701
Code : 72
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10673012
  • 204.00 € / Pack of 6
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Description
Description
XX4204709, XX4304701, XX4404703, 5614, and 00907P are replacement parts for Filter Holder XX4404700
SS Filter, Depends on filter type used.
Specifications
Specifications
Not Autoclavable
47 mm high Pressure SS filter holder
Non-sterile
O-Ring
Silicone