missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories for Sterifil™ Aseptic Systems

Used for general filtration applications and filtration of samples for particulate or biological contamination analysis

Brand:  Merck XX1104702

Code : 72

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10019810

  • 20.05 € / Each

Estimated Shipment: 15-07-2022
to see stock.
Add to basket

Description

Description

MilliporeSigma Polypropylene Support Screen, For Use With: For Swinnex 47, Surface Area: Support Screen, 47mm dia. Filter, Polypropylene, for Swinnex 47

For use with Sterifil™ Aseptic Systems
Specifications

Specifications

Not Autoclavable
Millipore Sterifil 47 mm aseptic vacuum Filter system
Non-sterile
47 mm
Base Holder with Support Screen
Polypropylene Base and Support Screen
Laboratory Filter Holders or Cyclones
Videos
SDS
Documents
Certificates
Special Offers

Special Offers