Learn More
Merck Two-Way Vacuum Valve
Accessory to convert a standard syringe into a hand vacuum pump
Brand: Merck XKEM00104
Code : 72
Additional Details : Weight : 25.00000kg
Description
- Made of polycarbonate
Specifications
Two-Way Valve | |
Polypropylene Pump | |
Polycarbonate |