missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Accessories for ZHE Hazardous Waste System
Accessories for filter holders for sample preparation.
108.00 € - 1115.00 €
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
10768051
|
Merck
XX2702512 |
2-Way Valve, 1/8 in. NPTM |
575.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10082143
|
Merck
XX6700010 |
Cylinder O-Rings |
335.00 €
Pack of 3 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10203451
|
Merck
YT3009003 |
Piston O-Rings, Wiper Seal |
188.00 €
Pack of 3 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10718441
|
Merck
YT3009002 |
Fitting Luer 1/8 in. NPTF |
198.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10430683
|
Merck
YT3009015 |
ZHE Tight Syringe, 50 mL |
1115.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10079890
|
Merck
YY2209068 |
O-Ring, 2-235, 90 mm, Fluoroelastomer |
308.00 €
Pack of 6 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10508411
|
Merck
YY2209064 |
Support Screen 90 mm |
214.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10708251
|
Merck
YT3009013 |
Quick-Disconnect Female Swagelok |
236.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10101451
|
Merck
YT3009001 |
Piston with O-Rings, Wiper Seal |
414.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10601872
|
Merck
YT3009016 |
Tedlar Bag |
1100.00 €
Pack of 10 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10602062
|
Merck
YT3009017 |
Tedlar Bag Adaptor |
424.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10661672
|
Merck
XX6700024 |
Vent/Relief Valve |
442.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10718251
|
Merck
YT3009014 |
Quick-Disconnect Male Swagelok |
108.00 €
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
Specifications
Not Autoclavable | |
Non-sterile |