missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Endothelin-1 Quantikine ELISA Kit

Product Code. 18731934
Click to view available options
Quantity:
1 Kit
1 Pack
Sufficient For:
1 x 96-well plate
6 x 96-well plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18731934

Brand: R&D Systems™ DET100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Endothelin-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Endothelin-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies multi-species Endothelin-1 in cell culture supernates (75 ul), serum (75 ul), edta plasma (75 ul), heparin plasma (75 ul), urine (75 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.39 - 25pg/mL
Assay Sensitivity 0.207pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine
Specificity Synthetic and Natural Endothelin-1
Sufficient For 1 x 96-well plate
Target Species Multi-species
For Use With (Application) ELISA
Quantity 1 Kit
Sample Volume Cell Culture Supernates (75uL),Serum (75uL),EDTA Plasma (75uL),Heparin Plasma (75uL),Urine (75uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ARCND3, EDN1, endothelin 1, endothelin-1, ET1, HDLCQ7, PPET1, Preproendothelin-1, QME
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.