Learn More
Invitrogen™ ENOX1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA597708
Product Code. 16500542
  • 464.00 € / 100µg
Description
ENOX1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Probably acts as a terminal oxidase of plasma electron transport from cytosolic NAD(P)H via hydroquinones to acceptors at the cell surface. Hydroquinone oxidase activity alternates with a protein disulfide-thiol interchange/oxidoreductase activity which may control physical membrane displacements associated with vesicle budding or cell enlargement. The activities oscillate with a period length of 24 minutes and play a role in control of the ultradian cellular biological clock.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ENOX1
Polyclonal
Unconjugated
ENOX1
bA64J21.1; candidate growth-related and time keeping constitutive hydroquinone (NADH) oxidase; candidate growth-related and time keeping constitutive hydroquinone [NADH] oxidase; cCNOX; Cell proliferation-inducing gene 38 protein; CNOX; Constitutive Ecto-NOX; ecto-NOX disulfide-thiol exchanger 1; ENOX1; Hydroquinone [NADH] oxidase; PIG38; Protein disulfide-thiol oxidoreductase
Rabbit
Protein G
RUO
55068
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
2.5 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q8TC92
ENOX1
Recombinant Human Ecto-NOX disulfide-thiol exchanger 1 protein (101-340AA).