missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ ep Dualfilter T.I.P.S.™ Filter Tips

Protects pipette and sample against unintentional and unnoticed contamination by aerosols, drops and splashes. Eppendorf™ ep Dualfilter T.I.P.S.™ Filter Tips has two-phase filter for contamination protection.

POA

Specifications

Certifications/Compliance EN 1822
For Use With (Application) DNA, RNA and Protein applications
Format Rack
View More Specs

Description

Description

  • Two filter layers
  • Provides dual protection against aerosols and biomolecules to guard both pipette and sample
  • Free of PCR inhibiting additives
  • Provide reliable downstream results
  • Provides high purity and quality

Applications

  • DNA applications (e.g. PCR)
  • RNA applications (e.g. Gen expression analysis)
  • Protein applications (e.g. Antibody Research)
  • Cell Culture applications (e.g. Media)
  • Applications with radio actives
  • All applications which use aerosol binding liquids
Specifications

Specifications

EN 1822
DNA, RNA and Protein applications
Rack
Videos
SDS
Documents

Documents

Product Certifications