missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epcam / CD326 Rabbit, Clone: EGP40/1555R, enQuireBio™
Rabbit Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | Epcam / CD326 |
---|---|
Clone | EGP40/1555R |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15930438
|
EnquireBio
4072-RBM15-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15920438
|
EnquireBio
4072-RBM15-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Epcam / CD326 | |
200μg/mL | |
Monoclonal | |
Rabbit | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
4072 | |
A synthetic peptide (around aa 20-60) from the N-terminus of human EPCAM protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
40-43kDa |
EGP40/1555R | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_002345 | |
EPCAM | |
IgG κ | |
Protein A/G Purified | |
Cell surface | |
HT29 cells or breast tumor |