Learn More
(-)-Epinephrine (+)-bitartrate salt, 98+%, ACROS Organics™
(-)-Epinephrine (+)-bitartrate salt, 98+%, C9H13NO3.C4H6O6, CAS Number-51-42-3, L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate, 25g, 100.0, 98.0, White to Yellow, 200-097-1, 333.29, GHS Signal Word: Danger
Brand: Acros Organics 430140250
Code : 29
Additional Details : CAS Number : 51-42-3 Weight : 0.07500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : I
Packaging | Glass bottle |
---|---|
Quantity | 25g |
Description
(-)-Epinephrine (+)-bitartrate salt, 98+%, C9H13NO3.C4H6O6, CAS Number-51-42-3, L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate, 25g, 100.0, 98.0, White to Yellow, 200-097-1, 333.29, GHS Signal Word: Danger
Chemical Identifiers
51-42-3 | |
333.29 | |
L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate | |
(2R,3R)-2,3-dihydroxybutanedioic acid;4-[(1S)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol |
C9H13NO3·C4H6O6 | |
YLXIPWWIOISBDD-KZKMUFAMSA-N | |
124080922 | |
CNCC(C1=CC(=C(C=C1)O)O)O.C(C(C(=O)O)O)(C(=O)O)O |
Specifications
(-)-Epinephrine (+)-bitartrate salt | |
0.5% max. | |
-20° to -24° (20°C, 589nm) (c=4, 1M HCl) | |
155.0°C | |
51-42-3 | |
100.0 | |
C9H13NO3·C4H6O6 | |
L-Adrenaline (+)-bitartrate salt, L-Adrenaline D-hydrogentartrate | |
CNCC(C1=CC(=C(C=C1)O)O)O.C(C(C(=O)O)O)(C(=O)O)O | |
333.29 | |
333.29 | |
98+% |
Authentic | |
Glass bottle | |
White to Yellow | |
25g | |
98.0 | |
98+% | |
15,3674 | |
YLXIPWWIOISBDD-KZKMUFAMSA-N | |
(2R,3R)-2,3-dihydroxybutanedioic acid;4-[(1S)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol | |
124080922 | |
Fine Crystalline Powder |
Safety and Handling
- (-)-Epinephrine (+)-bitartrate salt
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H300-Fatal if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.