Learn More
Eppendorf™ A-4-38 Rotor Adapters
For round and rectangular buckets
99.05 € - 104.00 €
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15244546
|
Eppendorf™
5702763004 |
9 x 5 mL Tubes |
104.00 €
Pack of 2 |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15138354
|
Eppendorf™
5702733008 |
1 x 5 mL Conical Tube |
99.05 €
Pack of 2 |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For use with Centrifuge 5702/ 5702 R/5702 RH and A-4-38 Rotor
- Designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs.
Specifications
Rotor Adapter | |
A-4-38 |