missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ μCuvette™ G1.0 and BioSpectrometer™ Fluorescence
Eppendorf™ μCuvette™ G1.0 and BioSpectrometer™ Fluorescence
Brand: Eppendorf™ 6137000901
Additional Details : Weight : 5.40000kg
Description
- Preprogrammed and freely programmable fluorescence methods
- Preprogrammed (modified) methods for Qubit DNA quantification kits (SS, HS, BR)
- Fluorescence measurement concentration of dsDNA down to 1 pg/µL, using Pico Green™
- Fluorescence measurement concentration of BSA down to 0.01 ng/µL
- Fluorescence excitation wavelength 470 nm, emission wavelengths 520 nm and 560 nm
- Fluorescence intensity across a range of 0.5 nM to 2,000 nM fluorescein
- Single consumable (UVette) for UV/Vis and fluorescence assays
Specifications
CE | |
Photometer | |
Eppendorf μCuvette™ G1.0 |
BioSpectrometer™ | |
230 V, 50/60 Hz | |
24 Months |