Learn More
Eppendorf™ S-4-72 Rotor Adapters
Available in a variety of configurations to suite a wide range of applications.
214.00 € - 225.00 €
Specifications
Type | Adapter |
---|---|
For Use With (Equipment) | S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
13256589
|
Eppendorf™
5804783000 |
8 x 15 mL Conical Tubes |
214.00 €
Pack of 2 |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13266589
|
Eppendorf™
5804784006 |
4 x 50 mL Conical Tubes |
214.00 €
Pack of 2 |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13276589
|
Eppendorf™
5804787005 |
1 x 250 mL Tubes |
214.00 €
Pack of 2 |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13286589
|
Eppendorf™
5804785002 |
2 x 50 mL Conical Tubes |
225.00 €
Pack of 2 |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12806223
|
Eppendorf™
5804789008 |
14 x 2.6/8 mL Tubes |
225.00 €
Pack of 2 |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13206599
|
Eppendorf™
5804791002 |
13 x 5.5/12 mL Tubes |
225.00 €
Pack of 2 |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13216599
|
Eppendorf™
5804792009 |
12 x 7/17 mL Tubes |
225.00 €
Pack of 2 |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15108364
|
Eppendorf™
5804793005 |
8 x 5 mL Conical Tubes |
225.00 €
Pack of 2 |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13226599
|
Eppendorf™
5804794001 |
26 x 1.5/2 mL Tubes |
225.00 €
Pack of 2 |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For use with Centrifuge 5804/5804 R and 5810/5810 R
Specifications
Adapter | |
S-4-72 (4 x 250mL) rotor in Model 5804/5810 centrifuges |