missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ twin.tec™ 96-Well Microbiology PCR Plates
Sterile, individually blistered, for all applications that need ultimate purity or certified absence of bacterial DNA
92.85 €
Specifications
Certifications/Compliance | Human DNA, DNase, RNase-free |
---|---|
Format | Standard |
Array | 8 x 12 |
Autoclavable | Autoclavable |
Material | Polypropylene |
Product Code | Brand | Color | Volume (Metric) Well | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Volume (Metric) Well | Price | Quantity & Availability | ||||
10492868
|
Eppendorf™
0030129300 |
Clear | 150 μL |
92.85 €
Pack of 10 |
Estimated Shipment: 26-04-2024
Log in to see stock. |
||||
10401728
|
Eppendorf™
0030129318 |
Blue | 150 μL |
92.85 €
Pack of 10 |
Estimated Shipment: 26-04-2024
Log in to see stock. |
||||
10176287
|
Eppendorf™
0030129326 |
Clear | 250 μL |
92.85 €
Pack of 10 |
Estimated Shipment: 26-04-2024
Log in to see stock. |
||||
10319597
|
Eppendorf™
0030129334 |
Blue | 250 μL |
92.85 €
Pack of 10 |
Estimated Shipment: 26-04-2024
Log in to see stock. |
||||
Description
- Thin-walled for optimum heat transfer to the sample
- Solid and torque-resistant
- Improved well-to-well tolerance
- Minimal binding of DNA, RNA and enzymes
- Cut-away corner and alphanumeric grid referencing
- Raised well rims for effective sealing; also reduces risk of cross-contamination
- Autoclavable (121°C, 20 min.)
- Certified to be free of any detectable human DNA, DNase, RNase and PCR inhibitors†
Skirted Plates
- Compatible with automated systems
- Skirted for label or bar code
- Fits most thermal cyclers and the MegaBACE™ capillary sequencer
- Within SBS footprint recommendations (127.76 ×85.47mm ±0.25mm)
- Stackable
- Eight holes in the skirt aid plate positioning and removal from the thermal cycler block
- Low-profile design enhances efficiency of PCR and enables the highest efficiency for small sample volumes
- 150μL max. well volume (when used with cap strips)
- Ideal for quantitative real-time PCR
- Fits most thermal cyclers
- 250μL max. well volume (when used with cap strips)
† Certificate, test procedures and detailed information available upon request.
Specifications
Human DNA, DNase, RNase-free | |
8 x 12 | |
Polypropylene | |
Sterile | |
10/Pk. | |
Clear |
Standard | |
Autoclavable | |
96 | |
RT-PCR applications | |
Yes |