Learn More
Eppendorf™ epTIPS™ Low-retention Pipettor Tips
Increase reproducibility and reduce the loss of expensive reagents in sensitive PCR and real-time PCR applications with these pipettor tips. Eppendorf™ epTIPS™ Low-retention Pipettor Tips are ideal for cell culture (media) applications and applications with samples that contain detergents.
Brand: Eppendorf™ 0030010035
Code : 96
Additional Details : Weight : 0.13500kg
Description
Increase reproducibility and reduce the loss of expensive reagents in sensitive PCR and real-time PCR applications with these pipettor tips. Eppendorf™ epTIPS™ Low-retention Pipettor Tips are ideal for cell culture (media) applications and applications with samples that contain detergents.
- Ultrahomogenous surface for maximum reproducibility
- Ultrahydrophobic surface for minimal sample loss
- Significantly reduced foam formation during pipetting
- Extreme resistance to chemicals
- Available in PCR clean and Eppendorf Quality
- Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile (sterile and pyrogen-free)
- No coating
Specifications
2 to 200 μL | |
Sterile | |
5.3cm | |
Low Retention | |
epT.I.P.S. |
Non-Filtered | |
Yellow | |
They satisfy the most stringent demands in medicine, the pharmaceutical and food industries, PCR*, molecular biology and cytotechnology | |
Eppendorf Pipetter |