missing translation for 'onlineSavingsMsg'
Learn More

Estriol, 97%, Thermo Scientific Chemicals

Catalog Number p-7077841
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
50-27-1
C18H24O3
288.38
PROQIPRRNZUXQM-ZXXIGWHRSA-N
estriol, oestriol, estratriol, ovestin, ovestrion, trihydroxyestrin, destriol, tridestrin, aacifemine, oestratriol
5756
CHEBI:27974
(8R,9S,13S,14S,16R,17R)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,16,17-triol
CC12CCC3C(C1CC(C2O)O)CCC4=C3C=CC(=C4)O
This item is not returnable. View return policy
50-27-1
C18H24O3
288.38
PROQIPRRNZUXQM-ZXXIGWHRSA-N
estriol, oestriol, estratriol, ovestin, ovestrion, trihydroxyestrin, destriol, tridestrin, aacifemine, oestratriol
5756
CHEBI:27974
(8R,9S,13S,14S,16R,17R)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,16,17-triol
CC12CCC3C(C1CC(C2O)O)CCC4=C3C=CC(=C4)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 280.0°C to 282.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 288.38
Percent Purity 97%
Chemical Name or Material Estriol
compliance-icons
Product Identifier
  • Estriol
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360-May damage fertility or the unborn child.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'