Learn More
Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
C2H6O, CAS Number-64-17-5, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702, CCO
Brand: Fisher Chemical E/0650DF/C17
Code : NEW
Additional Details : CAS Number : 64-17-5 Weight : 1.97250kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Packaging | HDPE plastic bottle |
---|---|
Quantity | 2.5L |
Description
C2H6O, CAS Number-64-17-5, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702, CCO
Chemical Identifiers
64-17-5 | |
46.069 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
CHEBI:16236 | |
CCO |
C2H6O | |
3568 | |
702 | |
ethanol |
Specifications
64-17-5 | |
3568 | |
CCO | |
46.069 | |
CHEBI:16236 | |
Liquid | |
7 | |
78°C | |
2.5L |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46.07g/mol | |
HDPE plastic bottle | |
-114°C | |
Colorless |
Safety and Handling
- Ethanol 96%, technical, duty free
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.