missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
57.42 EUR valid until 2024-12-31
Use promo code "21953" to get your promotional price.
57.42 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
64.78 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "21953" to get your promotional price.
57.42 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
64.78 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Amber glass bottle
HDPE plastic bottle
Metal can
Plastic Coated Glass bottle
Plastic container
Description
C2H6O, CAS Number-64-17-5, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702, CCO
Specifications
Packaging | Amber glass bottle |
Quantity | 500 mL |
Color | Colorless |
Formula Weight | 46.07g/mol |
pH | 7 |
Melting Point | -114°C |
Boiling Point | 78°C |
Physical Form | Liquid |
Safety and Handling
Product Identifier
- Ethanol, absolute
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P337+P313-If eye irritation persists: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'