missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, 99.8%, for HPLC, absolute, Thermo Scientific Chemicals
114.25 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Use promo code "21741" to get your promotional price.
Click to view available options
Quantity:
1 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS | 64-17-5, 7732-18-5 |
Quantity | 1 L |
Molecular Formula | C2H6O |
Linear Formula | CH3CH2OH |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
SMILES | CCO |
IUPAC Name | ethanol |
Molecular Weight (g/mol) | 46.069 |
PubChem CID | 702 |
Show More |
Safety and Handling
Product Identifier
- (±)-2-(2-Pyrrolidinyl)pyridine
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
- P337+P313-If eye irritation persists: Get medical advice/attention.
For Manufacturing and Laboratory Use. Not for beverage use
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'