missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Specifications
Specifications
| Chemical Name or Material | Ethanol |
| CAS | 64-17-5 |
| Analyte or Component Names | Ethanol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C2 H6 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Ethanol, Dehydrated Alcohol, Ethanol (9CI, ACI), Ethyl alcohol (6CI, 7CI, 8CI), 100C.NPA, Adechiru 750, AHD 2000, Alcare Hand Degermer, Alcohol, Alcohol anhydrous, Algrain, Alpet E, Alpet NV, Amanol SD, Anhydrol, Anhydrol PM 4085, Bioethanol, Black Warrant, CDA 19, CDA 19-200, Denatured alcohol, Denatured ethanol, Desinfektol EL, Duplicating Fluid 100C.NPA, Esumiru WK 88, Ethicap, Ethyl hydrate, Ethyl hydroxide, Germ-X, Hinetoless, ICON Dry, IMS 99, Infinity Pure, Jaysol, Jaysol S, LEK 01, Lux, Methylcarbinol, Molasses alcohol, Neocol CQ, NSC 85228, PM 6193-200, Potato alcohol, Prethanol A, SD 3A, SDA 23A, SDA 3A, Sekundasprit, Shunkol B, Sterillium Rub, Surfin 96, SY Fresh M, Synasol, Tecsol, Tecsol C, Traceable 95, Vinic alcohol |
| InChI Formula | InChI=1S/C2H6O/c1-2-3/h3H,2H2,1H3 |
| Solution Type | Neat |
| SMILES | CCO |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction