missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminobenzo[b]thiophene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 15476957
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15476957

Brand: Thermo Scientific Alfa Aesar H34050.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34761-09-6
Molecular Formula C11H11NO2S
Molecular Weight (g/mol) 221.27
MDL Number MFCD00661903
InChI Key XZSFZYVMRZLUOJ-UHFFFAOYSA-N
Synonym ethyl 3-aminobenzo b thiophene-2-carboxylate, ethyl 3-aminobenzothiophene-2-carboxylate, benzo b thiophene-2-carboxylic acid, 3-amino-, ethyl ester, 3-amino-benzo b thiophene-2-carboxylic acid ethyl ester, 3-aminobenzo b thiophene-2-carboxylic acid ethyl ester, maybridge1_003360, 3-amino-2-carboethoxy-benzothiophene, benzo b thiophene-2-carboxylicacid, 3-amino-, ethyl ester
PubChem CID 731510
IUPAC Name ethyl 3-amino-1-benzothiophene-2-carboxylate
SMILES CCOC(=O)C1=C(N)C2=CC=CC=C2S1

Specifications

Melting Point ∼82°C (sublimation)
Quantity 5 g
Formula Weight 221.28
Percent Purity 97%
Chemical Name or Material Ethyl 3-aminobenzo[b]thiophene-2-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt