Learn More
Ethyl 4-chloroacetoacetate, 98%, ACROS Organics™
Brand: Acros Organics 177640500
Code : 29
Additional Details : CAS Number : 638-07-3 Weight : 0.10000kg Transport : UN number : 2927 Chem class : 6.1 Pack group : II
Packaging | Glass bottle |
---|---|
Quantity | 50g |
Chemical Identifiers
638-07-3 | |
164.59 | |
OHLRLMWUFVDREV-UHFFFAOYSA-N | |
69484 | |
CCOC(=O)CC(=O)CCl |
C6H9ClO3 | |
MFCD00000939 | |
ethyl 4-chloroacetoacetate, 4-chloroacetoacetic acid ethyl ester, butanoic acid, 4-chloro-3-oxo-, ethyl ester, ethyl gamma-chloroacetoacetate, unii-5ae82250cm, ccris 6791, ethyl chloroacetyl acetate, butanoic acid, 4-chloro-3-oxo-ethylester, ethyl 4-chloro-3-oxobutyrate, ethyl 4-chloro-3-oxo-butanoate | |
ethyl 4-chloro-3-oxobutanoate |
Specifications
Ethyl 4-chloroacetoacetate | |
96°C | |
Glass bottle | |
1.21 | |
Colorless to Red-Yellow | |
50g | |
638-07-3 | |
C6H9ClO3 | |
MFCD00000939 | |
ethyl 4-chloroacetoacetate, 4-chloroacetoacetic acid ethyl ester, butanoic acid, 4-chloro-3-oxo-, ethyl ester, ethyl gamma-chloroacetoacetate, unii-5ae82250cm, ccris 6791, ethyl chloroacetyl acetate, butanoic acid, 4-chloro-3-oxo-ethylester, ethyl 4-chloro-3-oxobutyrate, ethyl 4-chloro-3-oxo-butanoate | |
OHLRLMWUFVDREV-UHFFFAOYSA-N | |
ethyl 4-chloro-3-oxobutanoate | |
69484 | |
Liquid |
1.2100g/mL | |
Authentic | |
1.4510 to 1.4530 | |
103.0°C (16.0 mmHg) | |
-8.0°C | |
98% | |
97.5% min. (GC) | |
ClCH2COCH2CO2C2H5 | |
03,663 | |
Solubility in water: 47.5g/L water (20°C). Other solubilities: soluble in ether,benzene,chloroform,,methylenechloride and carbontetrachloride | |
CCOC(=O)CC(=O)CCl | |
164.59 | |
164.59 | |
98% |
Safety and Handling
- Ethyl 4-chloroacetoacetate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H301-Toxic if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.