missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-methyloxazole-5-carboxylate, 97%, Thermo Scientific™

Catalog Number p-7077199
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
20485-39-6
C7H9NO3
155.15
MFCD00062573
XNMORZSEENWFLI-UHFFFAOYSA-N
88558
ethyl 4-methyl-1,3-oxazole-5-carboxylate
CCOC(=O)C1=C(C)N=CO1
This item is not returnable. View return policy
20485-39-6
C7H9NO3
155.15
MFCD00062573
XNMORZSEENWFLI-UHFFFAOYSA-N
88558
ethyl 4-methyl-1,3-oxazole-5-carboxylate
CCOC(=O)C1=C(C)N=CO1
Melting Point 32.0°C to 38.0°C
Boiling Point 100.0°C to 103.0°C (21.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Synonym ethyl 4-methyloxazole-5-carboxylate, ethyl4-methyloxazole-5-carboxylate, 4-methyloxazole-5-carboxylic acid ethyl ester, 5-ethoxycarbonyl-4-methyloxazole, ethyl 4-methyl-5-oxazolecarboxylate, 5-oxazolecarboxylic acid, 4-methyl-, ethyl ester, 4-methyl-oxazole-5-carboxylic acid ethyl ester, 4-methyl-5-carbethoxyoxazole, ethyl 4-methyl-oxazole-5-carboxylate, ethyl-4-methyl-1,3-oxazole-5-carboxylate
Formula Weight 155.15
Percent Purity 97%
Chemical Name or Material Ethyl 4-methyloxazole-5-carboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 4-methyl-1,3-oxazole-5-carboxylate, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'